missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Trifluoro-2-(trifluoromethyl)-4-penten-2-ol 98.0+%, TCI America™

Catalog No. p-7137503
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
T29685G 5 g
T296825G 25 g
2 options

Catalog No. T29685G

Supplier: TCI America T29685G

Only null left

Chemical Identifiers

CAS 646-97-9
Molecular Formula C6H6F6O
Molecular Weight (g/mol) 208.10
MDL Number MFCD03092986
InChI Key VHSCQANAKTXZTG-UHFFFAOYSA-N
PubChem CID 328870
IUPAC Name 1,1,1-trifluoro-2-(trifluoromethyl)pent-4-en-2-ol
SMILES OC(CC=C)(C(F)(F)F)C(F)(F)F

Specifications

Color Yellow
Boiling Point 100°C
Quantity 5 g
UN Number 1993
Formula Weight 208.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,1-Trifluoro-2-(trifluoromethyl)-4-penten-2-ol