accessibility menu, dialog, popup

UserName

OHH2CFFFFFF

CAS RN 646-97-9

IUPAC Name: 1,1,1-trifluoro-2-(trifluoromethyl)pent-4-en-2-ol
Synonyms: 1,1,1-trifluoro-2-(trifluoromethyl)pent-4-en-2-ol
Molecular Weight (g/mol): 208.1
Molecular Formula: C6H6F6O
InChi Key: VHSCQANAKTXZTG-UHFFFAOYSA-N
SMILES: OC(CC=C)(C(F)(F)F)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results