missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Trifluoro-2-(trifluoromethyl)-4-penten-2-ol 98.0+%, TCI America™

Catalog No. T296825G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T296825G 25 g
T29685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T296825G Supplier TCI America Supplier No. T296825G

Chemical Identifiers

CAS 646-97-9
Molecular Formula C6H6F6O
Molecular Weight (g/mol) 208.10
MDL Number MFCD03092986
InChI Key VHSCQANAKTXZTG-UHFFFAOYSA-N
PubChem CID 328870
IUPAC Name 1,1,1-trifluoro-2-(trifluoromethyl)pent-4-en-2-ol
SMILES OC(CC=C)(C(F)(F)F)C(F)(F)F

Specifications

Color Yellow
Boiling Point 100°C
Quantity 25 g
UN Number 1993
Formula Weight 208.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,1-Trifluoro-2-(trifluoromethyl)-4-penten-2-ol