missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Trifluoro-2,4-pentanedione, 98%

CAS: 367-57-7 | C5H5F3O2 | 154.09 g/mol

Supplier:  Thermo Scientific Chemicals A1468214

 View more versions of this product

Catalog No. AAA1468214


Only null left
Explore available promotions

Description

Description

1,1,1-Trifluoro-2,4-pentanedione has been used as reagent in the preparation of 2-alkylcarbonyl and 2-benzoyl-3-trifluoromethylquinoxaline 1,4-di-N-oxide derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,1-Trifluoro-2,4-pentanedione has been used as reagent in the preparation of 2-alkylcarbonyl and 2-benzoyl-3-trifluoromethylquinoxaline 1,4-di-N-oxide derivatives.

Solubility
Soluble in water (slightly) and many organic solvents.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

1,1,1-Trifluoro-2,4-pentanedione
1.274
26°C (79°F)
1.389
25 g
1705177
Soluble in water (slightly) and many organic solvents.
CC(=O)CC(=O)C(F)(F)F
154.09
154.09
367-57-7
105°C to 107°C
C5H5F3O2
MFCD00000427
UN1224
1,1,1-trifluoro-2,4-pentanedione, trifluoroacetylacetone, 1,1,1-trifluoroacetylacetone, trifluoroacetyl acetone, acetyl trifluoroacetone, 2,4-pentanedione, 1,1,1-trifluoro, tfpd-h, unii-9n20a8g8sw, alpha,alpha,alpha-trifluoroacetylacetone, a,a,a-trifluoroacetylacetone
SHXHPUAKLCCLDV-UHFFFAOYSA-N
1,1,1-trifluoropentane-2,4-dione
73943
98%

Chemical Identifiers

367-57-7
154.09
SHXHPUAKLCCLDV-UHFFFAOYSA-N
73943
CC(=O)CC(=O)C(F)(F)F
C5H5F3O2
MFCD00000427
1,1,1-trifluoro-2,4-pentanedione, trifluoroacetylacetone, 1,1,1-trifluoroacetylacetone, trifluoroacetyl acetone, acetyl trifluoroacetone, 2,4-pentanedione, 1,1,1-trifluoro, tfpd-h, unii-9n20a8g8sw, alpha,alpha,alpha-trifluoroacetylacetone, a,a,a-trifluoroacetylacetone
1,1,1-trifluoropentane-2,4-dione
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c

H226-H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.

EINECSNumber : 206-698-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only