missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,5,5,5-Hexamethyl-3-[(trimethylsilyl)oxy]-3-vinyltrisiloxane 98.0+%, TCI America™

Catalog No. p-7131132
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H16135ML 5 mL
H161325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H16135ML Supplier TCI America Supplier No. H16135ML
Only null left

Chemical Identifiers

CAS 5356-84-3
Molecular Formula C11H30O3Si4
Molecular Weight (g/mol) 322.70
MDL Number MFCD00054159
InChI Key CHEFFAKKAFRMHG-UHFFFAOYSA-N
Synonym vinyl tris trimethylsiloxy silane, vinyltris trimethylsiloxy silane, 1,1,1,5,5,5-hexamethyl-3-trimethylsilyl oxy-3-vinyltrisiloxane, tris trimethylsiloxy vinyl silane, trisiloxane, 3-ethenyl-1,1,1,5,5,5-hexamethyl-3-trimethylsilyl oxy, 4-ethenyl-2,2,6,6-tetramethyl-4-trimethylsilyl oxy-3,5-dioxa-2,4,6-trisilaheptane
PubChem CID 79316
IUPAC Name 4-ethenyl-2,2,6,6-tetramethyl-4-[(trimethylsilyl)oxy]-3,5-dioxa-2,4,6-trisilaheptane
SMILES C[Si](C)(C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)C=C

Specifications

Color Colorless
Boiling Point 88°C
Quantity 5 mL
Formula Weight 322.70
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,1,5,5,5-Hexamethyl-3-[(trimethylsilyl)oxy]-3-vinyltrisiloxane