accessibility menu, dialog, popup

UserName

CH3SiCH3H3COSiOSiH3CCH3H3COSiCH3H3CCH3H2C

CAS RN 5356-84-3

IUPAC Name: 4-ethenyl-2,2,6,6-tetramethyl-4-[(trimethylsilyl)oxy]-3,5-dioxa-2,4,6-trisilaheptane
Synonyms: 4-ethenyl-2,2,6,6-tetramethyl-4-[(trimethylsilyl)oxy]-3,5-dioxa-2,4,6-trisilaheptane
Molecular Weight (g/mol): 322.7
Molecular Formula: C11H30O3Si4
InChi Key: CHEFFAKKAFRMHG-UHFFFAOYSA-N
SMILES: C[Si](C)(C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)C=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results