missing translation for 'onlineSavingsMsg'
Learn More

1,1'-[1,4-Phenylenebis(methylene)]bis(4,4'-bipyridinium) Bis(hexafluorophosphate) 98.0+%, TCI America™

Catalog No. P1407100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1407100MG 100 mg
P14071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1407100MG Supplier TCI America Supplier No. P1407100MG

Chemical Identifiers

CAS 108861-20-7
Molecular Formula C28H24F12N4P2
Molecular Weight (g/mol) 706.456
MDL Number MFCD02063876
InChI Key ZURJGCCXYLZWSO-UHFFFAOYSA-N
Synonym 1,1′C-(p-Xylylene)bis(4,4′C-bipyridinium) Bis(hexafluorophosphate)
PubChem CID 11061517
IUPAC Name 4-pyridin-4-yl-1-[[4-[(4-pyridin-4-ylpyridin-1-ium-1-yl)methyl]phenyl]methyl]pyridin-1-ium;dihexafluorophosphate
SMILES C1=CC(=CC=C1C[N+]2=CC=C(C=C2)C3=CC=NC=C3)C[N+]4=CC=C(C=C4)C5=CC=NC=C5.F[P-](F)(F)(F)(F)F.F[P-](F)(F)(F)(F)F

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 706.46
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-[1,4-Phenylenebis(methylene)]bis(4,4′-bipyridinium) Bis(hexafluorophosphate)