accessibility menu, dialog, popup

UserName

NNNNFPFFFFFFPFFFFF

CAS RN 108861-20-7

IUPAC Name: 1-{[4-({[4,4'-bipyridin]-1-ium-1-yl}methyl)phenyl]methyl}-[4,4'-bipyridin]-1-ium; bis(hexafluoro-λ⁵-phosphanuide)
Synonyms: 1-[(4-{[4,4'-bipyridin]-1-ium-1-ylmethyl}phenyl)methyl]-[4,4'-bipyridin]-1-ium dihexafluorophosphate
Molecular Weight (g/mol): 706.46
Molecular Formula: C28H24F12N4P2
InChi Key: ZURJGCCXYLZWSO-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.F[P-](F)(F)(F)(F)F.C(C1=CC=C(C[N+]2=CC=C(C=C2)C2=CC=NC=C2)C=C1)[N+]1=CC=C(C=C1)C1=CC=NC=C1
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)