missing translation for 'onlineSavingsMsg'
Learn More

1,1'-[1,4-Phenylenebis(methylene)]bis(1-pyridinium) Dibromide 98.0+%, TCI America™

Catalog No. P2141250MG
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2141250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P2141250MG Supplier TCI America Supplier No. P2141250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Laboratory reagent offered at ≥98.0% purity.

Chemical Identifiers

CAS 14208-10-7
Molecular Formula C18H18Br2N2
Molecular Weight (g/mol) 422.164
MDL Number MFCD00043749
InChI Key MMKBQSJLLGHVIM-UHFFFAOYSA-L
Synonym 1,1′-(p-Xylylene)bis(1-pyridinium) Dibromide, 1,1′-(p-Xylylene)bis(1-pyridinium bromide), DPX
PubChem CID 3036637
IUPAC Name 1-[[4-(pyridin-1-ium-1-ylmethyl)phenyl]methyl]pyridin-1-ium;dibromide
SMILES C1=CC=[N+](C=C1)CC2=CC=C(C=C2)C[N+]3=CC=CC=C3.[Br-].[Br-]

Specifications

Quantity 250 mg
Formula Weight 422.16
Percent Purity ≥98.0% (HPLC,T)
Grade Laboratory
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-[1,4-Phenylenebis(methylene)]bis(1-pyridinium) Dibromide
RTECSNumber UU6857800
TSCA No
Recommended Storage Refrigerator