accessibility menu, dialog, popup

UserName

BrBrNN

CAS RN 14208-10-7

IUPAC Name: 1-({4-[(pyridin-1-ium-1-yl)methyl]phenyl}methyl)pyridin-1-ium dibromide
Synonyms: 1-{[4-(pyridin-1-ium-1-ylmethyl)phenyl]methyl}pyridin-1-ium dibromide
Molecular Weight (g/mol): 422.16
Molecular Formula: C18H18Br2N2
InChi Key: MMKBQSJLLGHVIM-UHFFFAOYSA-L
SMILES: [Br-].[Br-].C(C1=CC=C(C[N+]2=CC=CC=C2)C=C1)[N+]1=CC=CC=C1
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results