missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,1,3-Tetrachloropropane 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | -35°C |
| Color | Yellow |
| Boiling Point | 159°C |
| Quantity | 25 g |
| UN Number | 1993 |
| Formula Weight | 181.87 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 1,1,1,3-Tetrachloropropane |
Safety and Handling
| EINECSNumber | (2)-0083 |
| RTECSNumber | TZ7000000 |
| TSCA | Yes |