accessibility menu, dialog, popup

UserName

CAS RN 1070-78-6

ClClClCl

CAS RN 1070-78-6

IUPAC Name: 1,1,1,3-tetrachloropropane
Synonyms: propane, 1,1,1,3-tetrachloro-
Molecular Weight (g/mol): 181.87
Molecular Formula: C3H4Cl4
InChi Key: UTACNSITJSJFHA-UHFFFAOYSA-N
SMILES: ClCCC(Cl)(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results