missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,5,5,5-Heptamethyl-3-[(trimethylsilyl)oxy]trisiloxane 97.0+%, TCI America™

Catalog No. H161225ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H161225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H161225ML Supplier TCI America Supplier No. H161225ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17928-28-8
Molecular Formula C10H30O3Si4
Molecular Weight (g/mol) 310.687
MDL Number MFCD00040007
InChI Key RGMZNZABJYWAEC-UHFFFAOYSA-N
Synonym Methyltris(trimethylsiloxy)silane
PubChem CID 28838
IUPAC Name trimethyl-[methyl-bis(trimethylsilyloxy)silyl]oxysilane
SMILES C[Si](C)(C)O[Si](C)(O[Si](C)(C)C)O[Si](C)(C)C

Specifications

Color Colorless
Boiling Point 190°C
Quantity 25 mL
Formula Weight 310.69
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,1,3,5,5,5-Heptamethyl-3-[(trimethylsilyl)oxy]trisiloxane