accessibility menu, dialog, popup

UserName

CAS RN 17928-28-8

IUPAC Name: 2,2,4,6,6-pentamethyl-4-[(trimethylsilyl)oxy]-3,5-dioxa-2,4,6-trisilaheptane
Synonyms: methyl trimethicone
Molecular Weight (g/mol): 310.69
Molecular Formula: C10H30O3Si4
InChi Key: RGMZNZABJYWAEC-UHFFFAOYSA-N
SMILES: C[Si](C)(C)O[Si](C)(O[Si](C)(C)C)O[Si](C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results