missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,5,5,5-Heptamethyl-3-(3-glycidyloxypropyl)trisiloxane 96.0+%, TCI America™

Catalog No. h12665g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12665G 5 g
H126625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H12665G Supplier TCI America Supplier No. H12665G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7422-52-8
Molecular Formula C13H32O4Si3
Molecular Weight (g/mol) 336.65
MDL Number MFCD00053926
InChI Key YSIQPJVFCSCUMU-UHFFFAOYSA-N
PubChem CID 62635
IUPAC Name trimethyl-[methyl-[3-(oxiran-2-ylmethoxy)propyl]-trimethylsilyloxysilyl]oxysilane
SMILES C[Si](C)(C)O[Si](C)(CCCOCC1CO1)O[Si](C)(C)C

Specifications

Color Colorless
Boiling Point 98°C
Quantity 5 g
Formula Weight 336.65
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,1,3,5,5,5-Heptamethyl-3-(3-glycidyloxypropyl)trisiloxane