accessibility menu, dialog, popup

UserName

CAS RN 7422-52-8

IUPAC Name: 2,2,4,6,6-pentamethyl-4-{3-[(oxiran-2-yl)methoxy]propyl}-3,5-dioxa-2,4,6-trisilaheptane
Synonyms: 2,2,4,6,6-pentamethyl-4-[3-(oxiran-2-ylmethoxy)propyl]-3,5-dioxa-2,4,6-trisilaheptane
Molecular Weight (g/mol): 336.65
Molecular Formula: C13H32O4Si3
InChi Key: YSIQPJVFCSCUMU-UHFFFAOYNA-N
SMILES: C[Si](C)(C)O[Si](C)(CCCOCC1CO1)O[Si](C)(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results