missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3-Pentamethyldisiloxane 95.0+%, TCI America™

Catalog No. p-7135168
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P15355ML 5 mL
P153525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P15355ML Supplier TCI America Supplier No. P15355ML
Only null left

Chemical Identifiers

CAS 1438-82-0
Molecular Formula C5H15OSi2
Molecular Weight (g/mol) 147.344
MDL Number MFCD00053658
InChI Key XUKFPAQLGOOCNJ-UHFFFAOYSA-N
Synonym pentamethyldisiloxane, disiloxane, pentamethyl, 1,1,1,3,3-pentamethyldisiloxane, dimethyl trimethylsilyloxy silicon, disiloxane, 1,1,1,3,3-pentamethyl, dimethyl trimethylsilyl oxy silyl, dimethylsilyl oxy trimethylsilane
PubChem CID 6327260
IUPAC Name dimethyl(trimethylsilyloxy)silicon
SMILES C[Si](C)O[Si](C)(C)C

Specifications

Color Colorless
Boiling Point 86°C
Quantity 5 mL
UN Number 1993
Formula Weight 148.35
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,1,3,3-Pentamethyldisiloxane
EINECSNumber (7)-0477
TSCA Yes