missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoroisopropyl p-Toluenesulfonate 98.0+%, TCI America™

Catalog No. H12815G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12815G 5 g
H128125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H12815G Supplier TCI America Supplier No. H12815G
Only null left

Chemical Identifiers

CAS 67674-48-0
Molecular Formula C10H8F6O3S
Molecular Weight (g/mol) 322.22
MDL Number MFCD00039262
InChI Key QGSVBFAODCJVIZ-UHFFFAOYSA-N
Synonym 1,1,1,3,3,3-hexafluoroisopropyl p-toluenesulfonate, 2-propanol, 1,1,1,3,3,3-hexafluoro-, 4-methylbenzenesulfonate, 2-propanol, 1,1,1,3,3,3-hexafluoro-, 2-4-methylbenzenesulfonate, 1,1,1,3,3,3-hexafluoroisopropyl 4-toluenesulphonate, 2,2,2-trifluoro-1-trifluoromethyl ethyl toluene-p-sulphonate, hexafluoroisopropyl 4-toluenesulfonate, 1,1,1,3,3,3-hexafluoroisopropyl tosylate, 1,1,1,3,3,3-hexafluoroisopropyl-4-toluensesulfonate, 2-propanol,1,1,3,3,3-hexafluoro-, 4-methylbenzenesulfonate, p-toluenesulfonic acid 1,1,1,3,3,3-hexafluoroisopropyl ester
PubChem CID 98916
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-yl 4-methylbenzene-1-sulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OC(C(F)(F)F)C(F)(F)F

Specifications

Melting Point 45°C
Color White-Yellow
Boiling Point 134°C
Quantity 5 g
Formula Weight 322.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,1,1,3,3,3-Hexafluoroisopropyl p-Toluenesulfonate