accessibility menu, dialog, popup

UserName

CH3SOOOFFFFFF

CAS RN 67674-48-0

IUPAC Name: 1,1,1,3,3,3-hexafluoropropan-2-yl 4-methylbenzene-1-sulfonate
Synonyms: 1,1,1,3,3,3-hexafluoropropan-2-yl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 322.22
Molecular Formula: C10H8F6O3S
InChi Key: QGSVBFAODCJVIZ-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OC(C(F)(F)F)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results