missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,1,3,3,3-Hexafluoroacetone hydrate, 98%, Thermo Scientific™
Chemical Identifiers
| CAS | 10057-27-9 |
|---|---|
| Molecular Formula | C3F6O·xH2O |
| Molecular Weight (g/mol) | 166.02 |
| InChI Key | SNZAEUWCEHDROX-UHFFFAOYSA-N |
| Synonym | hexafluoroacetone trihydrate, 1,1,1,3,3,3-hexafluoropropan-2-one trihydrate, 2-propanone, hexafluoro-, trihydrate, hfa trihydrate, acetone, hexafluoro-, trihydrate, 2-propanone, 1,1,1,3,3,3-hexafluoro-, trihydrate, perfluoroacetone trihydrate, hexafluoro-2-propanone trihydrate, hexafluoro-2-propanone trihydrate; perfluoroacetone trihydrate, hexafluoroacetonetrihydrate |
| PubChem CID | 36719 |
| IUPAC Name | 1,1,1,3,3,3-hexafluoropropan-2-one;trihydrate |
| SMILES | C(=O)(C(F)(F)F)C(F)(F)F.O.O.O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC312120250
|
N/A | 25g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 10057-27-9 | |
| 166.02 | |
| hexafluoroacetone trihydrate, 1,1,1,3,3,3-hexafluoropropan-2-one trihydrate, 2-propanone, hexafluoro-, trihydrate, hfa trihydrate, acetone, hexafluoro-, trihydrate, 2-propanone, 1,1,1,3,3,3-hexafluoro-, trihydrate, perfluoroacetone trihydrate, hexafluoro-2-propanone trihydrate, hexafluoro-2-propanone trihydrate; perfluoroacetone trihydrate, hexafluoroacetonetrihydrate | |
| 1,1,1,3,3,3-hexafluoropropan-2-one;trihydrate |
| C3F6O·xH2O | |
| SNZAEUWCEHDROX-UHFFFAOYSA-N | |
| 36719 | |
| C(=O)(C(F)(F)F)C(F)(F)F.O.O.O |
Specifications
| 10057-27-9 | |
| 105.0°C to 108.0°C | |
| 97.5% min. (GC) | |
| 1.58 | |
| Solubility in water: soluble. | |
| C(=O)(C(F)(F)F)C(F)(F)F.O.O.O | |
| 166.02 | |
| 166.02 | |
| 1,1,1,3,3,3-Hexafluoroacetone hydrate |
| 1.5800g/mL | |
| Authentic | |
| C3F6O·xH2O | |
| hexafluoroacetone trihydrate, 1,1,1,3,3,3-hexafluoropropan-2-one trihydrate, 2-propanone, hexafluoro-, trihydrate, hfa trihydrate, acetone, hexafluoro-, trihydrate, 2-propanone, 1,1,1,3,3,3-hexafluoro-, trihydrate, perfluoroacetone trihydrate, hexafluoro-2-propanone trihydrate, hexafluoro-2-propanone trihydrate; perfluoroacetone trihydrate, hexafluoroacetonetrihydrate | |
| SNZAEUWCEHDROX-UHFFFAOYSA-N | |
| 1,1,1,3,3,3-hexafluoropropan-2-one;trihydrate | |
| 36719 | |
| 98% |
Safety and Handling
GHS H Statement Toxic if swallowed. Fatal in contact with skin. Toxic if inhaled. Causes skin irritation. Causes serious eye damage. Suspected of damaging fertility. Suspected of damaging the unborn child.
GHS P Statement Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection. Avoid breathing dust/fume/gas/mist/vapors/spray.
GHS Signal Word: Danger
RUO – Research Use Only