missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoroacetone hydrate, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 312120250

Quantity 25g

 View more versions of this product

Catalog No. AC312120250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

10057-27-9
166.02
hexafluoroacetone trihydrate, 1,1,1,3,3,3-hexafluoropropan-2-one trihydrate, 2-propanone, hexafluoro-, trihydrate, hfa trihydrate, acetone, hexafluoro-, trihydrate, 2-propanone, 1,1,1,3,3,3-hexafluoro-, trihydrate, perfluoroacetone trihydrate, hexafluoro-2-propanone trihydrate, hexafluoro-2-propanone trihydrate; perfluoroacetone trihydrate, hexafluoroacetonetrihydrate
1,1,1,3,3,3-hexafluoropropan-2-one;trihydrate
C3F6O·xH2O
SNZAEUWCEHDROX-UHFFFAOYSA-N
36719
C(=O)(C(F)(F)F)C(F)(F)F.O.O.O

Specifications

1,1,1,3,3,3-Hexafluoroacetone hydrate
1.5800g/mL
Authentic
C3F6O·xH2O
1.58
Solubility in water: soluble.
C(=O)(C(F)(F)F)C(F)(F)F.O.O.O
166.02
166.02
10057-27-9
105.0°C to 108.0°C
97.5% min. (GC)
25g
hexafluoroacetone trihydrate, 1,1,1,3,3,3-hexafluoropropan-2-one trihydrate, 2-propanone, hexafluoro-, trihydrate, hfa trihydrate, acetone, hexafluoro-, trihydrate, 2-propanone, 1,1,1,3,3,3-hexafluoro-, trihydrate, perfluoroacetone trihydrate, hexafluoro-2-propanone trihydrate, hexafluoro-2-propanone trihydrate; perfluoroacetone trihydrate, hexafluoroacetonetrihydrate
SNZAEUWCEHDROX-UHFFFAOYSA-N
1,1,1,3,3,3-hexafluoropropan-2-one;trihydrate
36719
98%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement Toxic if swallowed. Fatal in contact with skin. Toxic if inhaled. Causes skin irritation. Causes serious eye damage. Suspected of damaging fertility. Suspected of damaging the unborn child.

GHS P Statement Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection. Avoid breathing dust/fume/gas/mist/vapors/spray.

GHS Signal Word: Danger

SDS
Documents

Documents

Promotions

Promotions