missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,2-Tetrafluoro-4-iodo-2-(trifluoromethyl)butane 98.0+%, TCI America™

Catalog No. T25835G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T25835G Supplier TCI America Supplier No. T25835G

Chemical Identifiers

CAS 99324-96-6
Molecular Formula C5H4F7I
Molecular Weight (g/mol) 323.98
MDL Number MFCD00042316
InChI Key NRVDKMYDLZBFEH-UHFFFAOYSA-N
Synonym 1,1,1,2-tetrafluoro-4-iodo-2-trifluoromethyl butane, 4-iodo-2-trifluoromethyl-1,1,1,2-tetrafluoro-butane, 4-iodo-2-trifluoromethyl-1,1,1,2-tetrafluorobutane, 1,1,1,2-tetrafluoro-2-trifluoromethyl-4-iodobutane, acmc-209scc, cf3 2cfch2ch2i, 3,4,4,4-tetrafluoro-3-trifluoromethyl-1-iodobutane, 1-iodo-3,4,4,4-tetrafluoro-3-trifluoromethylbutane, 4-iodo-2 trifluoromethyl-1,1,1,2-tetrafluorobutane
PubChem CID 629659
IUPAC Name 1,1,1,2-tetrafluoro-4-iodo-2-(trifluoromethyl)butane
SMILES C(CI)C(C(F)(F)F)(C(F)(F)F)F

Specifications

Color Yellow
Boiling Point 110°C
Quantity 5 g
Formula Weight 323.98
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,1,2-Tetrafluoro-4-iodo-2-(trifluoromethyl)butane