CAS RN 99324-96-6
IUPAC Name:
1,1,1,2-tetrafluoro-4-iodo-2-(trifluoromethyl)butane
Synonyms:
1,1,1,2-tetrafluoro-4-iodo-2-(trifluoromethyl)butane
Molecular Weight (g/mol):
323.98
Molecular Formula:
C5H4F7I
InChi Key:
NRVDKMYDLZBFEH-UHFFFAOYSA-N
SMILES:
FC(F)(F)C(F)(CCI)C(F)(F)F
1
–
1
of
1
results
1
1,1,1,2-Tetrafluoro-4-iodo-2-(trifluoromethyl)butane 98.0+%, TCI America™
CAS: 99324-96-6 Molecular Formula: C5H4F7I Molecular Weight (g/mol): 323.98 MDL Number: MFCD00042316 InChI Key: NRVDKMYDLZBFEH-UHFFFAOYSA-N Synonym: 1,1,1,2-tetrafluoro-4-iodo-2-trifluoromethyl butane,4-iodo-2-trifluoromethyl-1,1,1,2-tetrafluoro-butane,4-iodo-2-trifluoromethyl-1,1,1,2-tetrafluorobutane,1,1,1,2-tetrafluoro-2-trifluoromethyl-4-iodobutane,acmc-209scc,cf3 2cfch2ch2i,3,4,4,4-tetrafluoro-3-trifluoromethyl-1-iodobutane,1-iodo-3,4,4,4-tetrafluoro-3-trifluoromethylbutane,4-iodo-2 trifluoromethyl-1,1,1,2-tetrafluorobutane PubChem CID: 629659 IUPAC Name: 1,1,1,2-tetrafluoro-4-iodo-2-(trifluoromethyl)butane SMILES: C(CI)C(C(F)(F)F)(C(F)(F)F)F