missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,2,2,3,4,5,5,5-Decafluoro-3-methoxy-4-(trifluoromethyl)pentane 98.0+%, TCI America™

Catalog No. d448425g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
D448425G 25 g
D4484500G 500 g
2 options

Catalog No. D448425G

Supplier: TCI America D448425G

Only null left

Chemical Identifiers

CAS 132182-92-4
Molecular Formula C7H3F13O
Molecular Weight (g/mol) 350.079
MDL Number MFCD07784227
InChI Key QKAGYSDHEJITFV-UHFFFAOYSA-N
PubChem CID 14689855
IUPAC Name 1,1,1,2,2,3,4,5,5,5-decafluoro-3-methoxy-4-(trifluoromethyl)pentane
SMILES COC(C(C(F)(F)F)(C(F)(F)F)F)(C(C(F)(F)F)(F)F)F

Specifications

Color Colorless
Boiling Point 100°C
Quantity 25 g
Formula Weight 350.08
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,1,2,2,3,4,5,5,5-Decafluoro-3-methoxy-4-(trifluoromethyl)pentane
EINECSNumber (2)-4055
TSCA Yes