accessibility menu, dialog, popup

UserName

CH3OFFFFFFFFFFFFF

CAS RN 132182-92-4

IUPAC Name: 1,1,1,2,2,3,4,5,5,5-decafluoro-3-methoxy-4-(trifluoromethyl)pentane
Synonyms: 1,1,1,2,2,3,4,5,5,5-decafluoro-3-methoxy-4-(trifluoromethyl)pentane
Molecular Weight (g/mol): 350.08
Molecular Formula: C7H3F13O
InChi Key: QKAGYSDHEJITFV-UHFFFAOYNA-N
SMILES: COC(F)(C(F)(F)C(F)(F)F)C(F)(C(F)(F)F)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results