missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One 0.5 mL Polypropylene Microcentrifuge Tubes
Description
- Material: Polypropylene
- For single use only
- With attached cap
- Free of detectable DNase/RNase, human DNA and pyrogens
- Non-Pyrogenic
- High chemical and temperature resistance
- Suitable for Viatron centri
Specifications
Specifications
| Color | Natural |
| Material | Polypropylene |
| Volume (Metric) | 0.5mL |
| Sterility | Non-sterile |