missing translation for 'onlineSavingsMsg'
Learn More

0.1% Formic Acid, Aqueous for LC-MS, Reagents

Catalog No. 01335308
Change view
Click to view available options
Quantity:
500 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335308 4 L
01335307 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01335308 Supplier Labchem Inc Supplier No. CF0006004C
Only null left

0.1% Formic Acid, Aqueous for LC-MS, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 64-18-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Methanoic Acid, Formylic Acid, Aminic Acid
IUPAC Name water
SMILES O

Specifications

pH 2.71
Boiling Point 100°C
Color Colorless
Percent Purity 0.1%
Grade Reagent
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 1.0002
Chemical Name or Material 0.1% Formic Acid, Aqueous for LC-MS