missing translation for 'onlineSavingsMsg'
Learn More

0.07% n-Hexylamine in HPLC Grade Isopropanol, Reagents

Catalog No. 01334696
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334696 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01334696 Supplier Labchem Inc Supplier No. CN0005004C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

0.07% n-Hexylamine in HPLC Grade Isopropanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-63-0 , 111-26-2
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym n-Hexylamine, 1-Aminohexane
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Color Colorless
Boiling Point 82.5°C
Packaging Glass Bottle
Quantity 4 L
Specific Gravity 0.7854
Grade Reagent
Physical Form Liquid
Chemical Name or Material 0.07% n-Hexylamine in HPLC Grade Isopropanol