Get a Freeze Cube When You Buy Fisherbrand Cryotubes
When you buy one case of qualifying Fisherbrand™ Cryogenic Storage Tubes, you’ll get one Fisherbrand Controlled Freeze Cube at no additional cost.
Order using the catalog numbers below. When you’ve made your qualifying purchase, log in to the Redemption Center to redeem this offer.
Features and Benefits
Choose externally threaded Fisherbrand cryotubes with HDPE closures for aseptic handling or internally threaded tubes, featuring polypropylene closures with silicone gaskets for tight seals. Fisherbrand Controlled Freeze Cubes hold 12 x 1 to 2 mL tubes and achieve a cooling rate of –1℃/minute to support better cell preservation.
| Catalog Number | Description | ||
|---|---|---|---|
Fisherbrand Controlled Freeze Cube, Purple | Learn More | ||
Fisherbrand Controlled Freeze Cube, Green | Learn More | ||
Fisherbrand Controlled Freeze Cube, Blue | Learn More |