Get a Microvolume Plate with Spectrophotometer Purchase
New Lab Start-up Program
When you purchase a qualifying Agilent BioTek Epoch 2 Microplate Spectrophotometer, you’ll get an Agilent Take3 Microvolume Plate at no additional cost. Once you’ve made your qualifying purchase, log in to the Redemption Center to redeem this offer.
NOTE: Promotion may be used for up to two qualifying products purchased on the same order.
Features and Benefits
Measure the UV-Vis absorbance of samples in 6- to 384-well microplates, cuvettes, and more with Agilent BioTek Epoch 2 Microplate Spectrophotometers. An optional touchscreen interface makes choosing predefined protocols or defining custom workflows simple and easy.
Simultaneously measure multiple 2 ÎĽL samples without dilution or specialized equipment with the Agilent Take3 Microvolume Plate.
Get a Microvolume Plate with Spectrophotometer Purchase
Redeem Now
Promotion: 661
You must apply and qualify for NLSU to redeem this promotion.
Qualifying Products
    Catalog Number Description Price Quantity & Availability  
Success
BTEPOCH2NSC Details
GREENER_CHOICE
Agilent BioTek Epoch 2 Microplate Spectrophotometer Includes: Cuvette Port
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Product Specifications
Wavelength Range 200 to 999 nm, 1 nm increments
Includes Cuvette Port
Height (English) 22 in.
Measurement Speed 96 wells: 8 sec., 384 wells: 14 sec.
Model EPOCH2NSC
For Use With (Application) Nucleic acid and protein quantification to ELISA and microbial growth assays
Light Source Xenon Flash Lamp
Hertz 50/60 Hz
Certifications/Compliance CE and TUV marked. RoHS compliant.
Accuracy ±2 nm
Detector Type Photodiode
BTEPOCH2TSC Details
GREENER_CHOICE
Agilent BioTek Epoch 2 Microplate Spectrophotometer Includes: Touchscreen; Cuvette port
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Product Specifications
Wavelength Range 200 to 999 nm, 1 nm increments
Includes Touchscreen; Cuvette port
Height (English) 22 in.
Measurement Speed 96 wells: 8 sec., 384 wells: 14 sec.
Model EPOCH2TSC
For Use With (Application) Nucleic acid and protein quantification to ELISA and microbial growth assays
Light Source Xenon Flash Lamp
Hertz 50/60 Hz
Certifications/Compliance CE and TUV marked. RoHS compliant.
Accuracy ±2 nm
Detector Type Photodiode
BTEPOCH2TSI Details
GREENER_CHOICE
Agilent BioTek Epoch 2 Microplate Spectrophotometer, IVD Includes: Touchscreen
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Product Specifications
Wavelength Range 200 to 999 nm, 1 nm increments
Includes Touchscreen
Height (English) 22 in.
Measurement Speed 96 wells: 8 sec., 384 wells: 14 sec.
Model EPOCH2TS
For Use With (Application) Nucleic acid and protein quantification to ELISA and microbial growth assays
Light Source Xenon Flash Lamp
Hertz 50/60 Hz
Certifications/Compliance CE and TUV marked. RoHS compliant.
Accuracy ±2 nm
Detector Type Photodiode
BTEPOCH2NCI Details
GREENER_CHOICE
Agilent BioTek Epoch 2 Microplate Spectrophotometer, IVD Includes: Cuvette Port
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Product Specifications
Wavelength Range 200 to 999 nm, 1 nm increments
Includes Cuvette Port
Height (English) 22 in.
Measurement Speed 96 wells: 8 sec., 384 wells: 14 sec.
Model EPOCH2NSC
For Use With (Application) Nucleic acid and protein quantification to ELISA and microbial growth assays
Light Source Xenon Flash Lamp
Hertz 50/60 Hz
Certifications/Compliance CE and TUV marked. RoHS compliant.
Accuracy ±2 nm
Detector Type Photodiode
BTEPOCH2NSI Details
GREENER_CHOICE
Agilent BioTek Epoch 2 Microplate Spectrophotometer, IVD Includes: No Touchscreen, No Cuvette Port
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Product Specifications
Wavelength Range 200 to 999 nm, 1 nm increments
Includes No Touchscreen, No Cuvette Port
Height (English) 22 in.
Measurement Speed 96 wells: 8 sec., 384 wells: 14 sec.
Model EPOCH2NS
For Use With (Application) Nucleic acid and protein quantification to ELISA and microbial growth assays
Light Source Xenon Flash Lamp
Hertz 50/60 Hz
Certifications/Compliance CE and TUV marked. RoHS compliant.
Accuracy ±2 nm
Detector Type Photodiode
BTEPOCH2TCI Details
GREENER_CHOICE
Agilent BioTek Epoch 2 Microplate Spectrophotometer, IVD Includes: Touchscreen, Cuvette Port
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Product Specifications
Wavelength Range 200 to 999 nm, 1 nm increments
Includes Touchscreen, Cuvette Port
Height (English) 22 in.
Measurement Speed 96 wells: 8 sec., 384 wells: 14 sec.
Model EPOCH2TSC
For Use With (Application) Nucleic acid and protein quantification to ELISA and microbial growth assays
Light Source Xenon Flash Lamp
Hertz 50/60 Hz
Certifications/Compliance CE and TUV marked. RoHS compliant.
Accuracy ±2 nm
Detector Type Photodiode
BTEPOCH2TS Details
GREENER_CHOICE
Agilent BioTek Epoch 2 Microplate Spectrophotometer Includes: Touchscreen only
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Product Specifications
Wavelength Range 200 to 999 nm, 1 nm increments
Includes Touchscreen only
Height (English) 22 in.
Measurement Speed 96 wells: 8 sec., 384 wells: 14 sec.
Model EPOCH2TS
For Use With (Application) Nucleic acid and protein quantification to ELISA and microbial growth assays
Light Source Xenon Flash Lamp
Hertz 50/60 Hz
Certifications/Compliance CE and TUV marked. RoHS compliant.
Accuracy ±2 nm
Detector Type Photodiode
BTEPOCH2NS Details
GREENER_CHOICE
Agilent BioTek Epoch 2 Microplate Spectrophotometer Includes: No Touchscreen, No Cuvette Port
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Product Specifications
Wavelength Range 200 to 999 nm, 1 nm increments
Includes No Touchscreen, No Cuvette Port
Height (English) 22 in.
Measurement Speed 96 wells: 8 sec., 384 wells: 14 sec.
Model EPOCH2NS
For Use With (Application) Nucleic acid and protein quantification to ELISA and microbial growth assays
Light Source Xenon Flash Lamp
Hertz 50/60 Hz
Certifications/Compliance CE and TUV marked. RoHS compliant.
Accuracy ±2 nm
Detector Type Photodiode
Redeemable Product
Catalog NumberDescription
Catalog Number
Description
Agilent BioTek Take3 Microvolume Plate