accessibility menu, dialog, popup

UserName

CAS RN 29898-72-4

NHNN

CAS RN 29898-72-4

IUPAC Name: 4-(4,5-diphenyl-1H-imidazol-2-yl)benzonitrile
Synonyms: 4-(4,5-diphenyl-1H-imidazol-2-yl)benzonitrile
Molecular Weight (g/mol): 321.38
Molecular Formula: C22H15N3
InChi Key: KGOZFYOJDPVGMV-UHFFFAOYSA-N
SMILES: N#CC1=CC=C(C=C1)C1=NC(=C(N1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results