missing translation for 'onlineSavingsMsg'
Learn More

4-(4,5-Diphenyl-1H-imidazol-2-yl)benzonitrile 98.0+%, TCI America™

Catalog No. D4655200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4655200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4655200MG Supplier TCI America Supplier No. D4655200MG
Only null left

Chemical Identifiers

CAS 29898-72-4
Molecular Formula C22H15N3
Molecular Weight (g/mol) 321.383
InChI Key KGOZFYOJDPVGMV-UHFFFAOYSA-N
Synonym 2-(4-Cyanophenyl)-4,5-diphenylimidazole
PubChem CID 12099795
IUPAC Name 4-(4,5-diphenyl-1H-imidazol-2-yl)benzonitrile
SMILES C1=CC=C(C=C1)C2=C(N=C(N2)C3=CC=C(C=C3)C#N)C4=CC=CC=C4

Specifications

Melting Point 275°C
Color White-Yellow
Quantity 200 mg
Formula Weight 321.38
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4,5-Diphenyl-1H-imidazol-2-yl)benzonitrile