Get a Heating Block with a Fisherbrand Dry Bath Purchase
New Lab Start-up Program
When you buy a qualifying Fisherbrand™ Isotemp™ Digital Dry Bath, you’ll get a heating block in your choice of configuration at no additional cost. View qualifying and redeemable products below. After making your qualifying purchase, log in to the Redemption Center to redeem this offer.
Features and Benefits
Fisherbrand Isotemp Digital Dry Baths offer a range of configurations for interchangeable modular blocks to suit a variety of basic to advanced applications with a maximum temperature of 130°C. They include a timer and are available in one-, two-, and four-block sizes.
Get a Heating Block with a Fisherbrand Dry Bath Purchase
Redeem Now
Promotion: 223
You must apply and qualify for NLSU to redeem this promotion.
Qualifying Products
    Catalog Number Description Price Quantity & Availability  
Success
88860021 Details
Fisherbrand™ Isotemp™ Digital Dry Baths/Block Heaters No. of Places: 1
Product Specifications
Height (Metric) 100 mm
Height (English) 3.9 in.
No. of Places 1
Length (Metric) 288 mm
Temperature Accuracy ≤ ± 0.5°C
Type Dry Bath Block
Width (English) 7.9 in.
Warranty 2 Year, Replacement
Plug Type US
Temperature Range (Metric) Ambient +5° to 130°C
Material Powder-coated Steel
Fuse 250V 2.5A
Certifications/Compliance CE, cULus, RoHS
Weight (Metric) 2.9 kg
Timer 0 to 99:59 min or continuous
Weight (English) 6.39 lb.
Relative Humidity <80%
Includes Dry bath, US style plug, block lifter
Heat Up Time ≤ 20 min. 30° to 130°C
Electrical Requirements 100/120 V, 50/60 Hz
Temperature Uniformity ≤ ± 1°C
Voltage 100/120 V
Length (English) 11.3 in.
Dimensions (L x W x H) 11.3 x 7.9 x 3.9 in. (288 x 200 x 100 mm)
Frequency 50/60 Hz
Controller Type PID
Width (Metric) 200 mm
88860023 Details
Fisherbrand™ Isotemp™ Digital Dry Baths/Block Heaters No. of Places: 4
Product Specifications
Height (Metric) 100 mm
Height (English) 3.9 in.
No. of Places 4
Length (Metric) 450 mm
Temperature Accuracy ≤ ± 0.5°C
Type Dry Bath Block
Width (English) 7.9 in.
Warranty 2 Year, Replacement
Plug Type US
Temperature Range (Metric) Ambient +5° to 130°C
Material Powder-coated Steel
Fuse 250V 2.5A
Certifications/Compliance CE, cULus, RoHS
Weight (Metric) 4.7 kg
Timer 0 to 99:59 min or continuous
Weight (English) 10.36 lb.
Relative Humidity <80%
Includes Dry bath, US style plug, block lifter
Heat Up Time ≤ 20 min. 30° to 130°C
Electrical Requirements 100/120 V, 50/60 Hz
Temperature Uniformity ≤ ± 1°C
Voltage 100/120 V
Length (English) 17.7 in.
Dimensions (L x W x H) 17.7 x 7.9 x 3.9 in. (450 x 200 x 100 mm)
Frequency 50/60 Hz
Controller Type PID
Width (Metric) 200 mm
88860022 Details
Fisherbrand™ Isotemp™ Digital Dry Baths/Block Heaters No. of Places: 2
Product Specifications
Height (Metric) 100 mm
Height (English) 3.9 in.
No. of Places 2
Length (Metric) 318 mm
Temperature Accuracy ≤ ± 0.5°C
Type Dry Bath Block
Width (English) 7.9 in.
Warranty 2 Year, Replacement
Plug Type US
Temperature Range (Metric) Ambient +5° to 130°C
Material Powder-coated Steel
Fuse 250V 5A
Certifications/Compliance CE, cULus, RoHS
Weight (Metric) 3.3 kg
Timer 0 to 99:59 min or continuous
Weight (English) 7.28 lb.
Relative Humidity <80%
Includes Dry bath, US style plug, block lifter
Heat Up Time ≤ 20 min. 30° to 130°C
Electrical Requirements 100/120 V, 50/60 Hz
Temperature Uniformity ≤ ± 1°C
Voltage 100/120 V
Length (English) 12.5 in.
Dimensions (L x W x H) 12.5 x 7.9 x 3.9 in. (318 x 200 x 100 mm)
Frequency 50/60 Hz
Controller Type PID
Width (Metric) 200 mm
Redeemable Products
Catalog NumberDescription
Catalog Number
Description
Fisherbrand Heating Block, 46 x 6 mm Diameter
Learn More
Catalog Number
Description
Fisherbrand Heating Block, 28 x 10 mm Diameter
Learn More
Catalog Number
Description
Fisherbrand Heating Block, 28 x 1.5 mL
Learn More
Catalog Number
Description
Fisherbrand Heating Block, 28 x 2.0 mL
Learn More
Catalog Number
Description
Fisherbrand Heating Block, 24 x 13 mm Diameter
Learn More
Catalog Number
Description
Fisherbrand Heating Block, 15 x 16 mm Diameter
Learn More
Catalog Number
Description
Fisherbrand Heating Block, 12 x 18 mm Diameter
Learn More
Catalog Number
Description
Fisherbrand Heating Block, 8 x 20 mm Diameter
Learn More
Catalog Number
Description
Fisherbrand Heating Block, 6 x 25 mm Diameter
Learn More
Catalog Number
Description
Fisherbrand Heating Block, 40 x 0.5 mL
Learn More
Catalog Number
Description
Fisherbrand Heating Block, ELISA, 1 x 96-Well
Learn More
Catalog Number
Description
Fisherbrand Heating Block, Mixed, 6 and 10 mm Diameter