Get an Extra Electrode with Thermo Scientific Meter Kit Purchase
When you buy a qualifying Thermo Scientific™ Orion Star™ meter kit, you’ll get an extra redeemable electrode at no additional cost. View qualifying and redeemable products below. After making your qualifying purchase, log in to the Redemption Center to redeem this offer.
The extra electrode will be the same as the one that comes in the original meter kit.
Features and Benefits
Record accurate and reliable pH, mV, ORP, and temperature measurements with the Thermo Scientific Orion Star A211 Benchtop pH Meter. This meter is ideal for a wide range of applications and advanced pH analysis in the lab. Perform up to a 5-point pH calibration and log up to 2,000 data point sets with time and date stamp. Use the included electrode stand to easily place sensors in samples and simplify storage to minimize breakage.
Catalog Number | Description | Price | Quantity & Availability |
---|
Catalog Number | Description | ||
---|---|---|---|
Thermo Scientific™ Orion™ ROSS Sure-Flow™ pH Electrodes, BNC Connector | Learn More | ||
Thermo Scientific™ Orion™ ROSS Ultra™ Glass Triode™ pH/ATC Combination Electrodes | Learn More | ||
Thermo Scientific Orion ROSS Ultra Refillable pH/ATC Triode Combination Electrodes | Learn More |