UserName
DWK Life Sciences WHEATON CompletePAK Vial Kits
DWK Life Sciences WHEATON CompletePAK Vial Kits
Kitted and Stand-Alone Options for Any Application
Explore new additions to the DWK Life Sciences™WHEATON™ vial collection that include sterile crimp top vials, rubber stoppers, and seals for your discovery and research, pharmaceutical compounding, aseptic injectables filling, freeze drying, and drug packaging.  
The DWK Life Sciences™WHEATON™ CompletePAK™ assortment now offers amber vials for light-sensitive contents and the ModPAK™ set is designed to meet your bulk packaging needs. Select an off-the-shelf option or ask us about custom configurations.
The glass vials are made from Type l borosilicate glass available in 2, 5, or 10mL vial sizes and packaged in sterile polypropylene trays. Stoppers come in Ultra Pure* or Omni Flex 3G** coated varieties (plug or igloo style) with red or blue flip seals. Components can also be purchased separately.
To ensure quality, all parts undergo particulate cleaning, depyrogenation, and sterilization. CompletePAK and ModPAK products are completely traceable and each kit includes certification documentation that they meet or exceed critical USP (United States Pharmacopeia) standards enforced by the FDA:
  • USP <788> Particulate Matter in Injections
  • USP <85> Bacterial Endotoxins Testing
  • USP <71> Sterility Testing 
  • Closure Container Integrity (CCI) Testing ─ Results available upon request 
*Ultra Pure stoppers are made from a gray bromobutyl rubber compound (46 Durometer) with a high level of chemical purity. They are free of latex, nitrosamines, and 2-mercaptobenzothiazole (2-MCBT), have a low gas permeability, and can be used for most lyophilization applications.
**Omni Flex 3G is a proprietary, flexible fluoropolymer spray coating applied to bromobutyl stoppers to create an inert barrier against organic molecules and metal ions.