Thermo Scientific Chemical Analysis Instrumentation
Rapid Materials Testing
Featured Products
Industry-Leading Expertise
Shorten your discovery paths, efficiently turn your data into answers, and conduct efficient quality control assessments with instruments for measurement control and sample identification, radiation detection and identification, and spectroscopy. Find durably constructed devices from small handheld meters to large portal monitors.
Pharmaceutical Production Support | Chemical and Narcotic Identification
Thermo Scientific TruNarc Narcotics Analyzer
Learn how the Thermo Scientific TruNarc handheld analyzer can help you safely detect and identify narcotics, precursors, and cutting agents.
Pharmaceutical Production Support
Real-Time Monitoring with PAT Software
Real-Time Monitoring with PAT Software
Use the Thermo Scientificâ„¢ MarqMetrixâ„¢ All-In-One Process Raman Analyzer in situations where time-to-results is key. Conduct real-time, non-destructive, and immediate analysis without sample preparation.
Shop MarqMetrix Analyzers
Rapid Material Confirmations
Rapid Material Confirmations
Confirm the composition of your raw materials and finished products using Raman spectroscopy with the Thermo Scientificâ„¢ TruScanâ„¢ RM. Light, fast, and portable, use TruScan at the point of need to minimize sampling costs and speed inventory turn-around.
Shop TruScan RM
Chemical and Narcotic Identification
Lab-Quality Results in the Palm of Your Hand
Lab-Quality Results in the Palm of Your Hand
The Thermo Scientificâ„¢ TruScan G3 Handheld Raman Analyzer is a game-changing solution for quality control in pharmaceutical and biotechnology manufacturing. With its lab-quality performance and rapid testing capabilities, this handheld device empowers non-technical operators to effortlessly perform raw material ID testing, eliminating the need for costly and time-consuming lab visits.
Shop TruScan Analyzers
Identify Narcotics and Precursors
Identify Narcotics and Precursors
Quickly detect narcotics, stimulants, depressants, hallucinogens, and analgesics with Thermo Scientificâ„¢ TruNarcâ„¢ Handheld Narcotics Analyzers. TruNarc uses Raman spectroscopy to identify more than 530 illicit and abused narcotics, fentanyl compounds, analogs, pharmaceutical variants, and NPP and ANPP fentanyl precursors at the point of need.
Shop TruNarc Analyzers
Fast, Accurate Chemical Identification
Fast, Accurate Chemical Identification
Simplify your work with the Thermo Scientificâ„¢ TruDefenderâ„¢ FTX Analyzer that offers rapid and accurate chemical identification using FTIR.
NOTE: Not sold in all countries. Contact your Fisher Scientific sales representative for availability information.
Shop TruDefender FTX Analyzers