accessibility menu, dialog, popup

UserName

CAS RN 99-29-6

NH2ClBrNOO

CAS RN 99-29-6

IUPAC Name: 2-bromo-6-chloro-4-nitroaniline
Synonyms: 2-bromo-6-chloro-4-nitroaniline
Molecular Weight (g/mol): 251.46
Molecular Formula: C6H4BrClN2O2
InChi Key: GLUCALKKMFBJEB-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C=C(C=C1Br)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results