accessibility menu, dialog, popup

UserName

OHNO(S)NH

CAS RN 96293-17-3

IUPAC Name: (2S)-2-[(2-benzoylphenyl)carbamoyl]-1-benzylpyrrolidin-1-ium
Synonyms: (2S)-2-[(2-benzoylphenyl)carbamoyl]-1-benzylpyrrolidin-1-ium
Molecular Weight (g/mol): 385.49
Molecular Formula: C25H25N2O2
InChi Key: IPSABLMEYFYEHS-QHCPKHFHSA-O
SMILES: O=C(NC1=CC=CC=C1C(=O)C1=CC=CC=C1)[C@@H]1CCC[NH+]1CC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results