accessibility menu, dialog, popup

UserName

CH3N(E/Z)

CAS RN 96184-40-6

IUPAC Name: 4-[4-(prop-1-en-1-yl)cyclohexyl]benzonitrile
Synonyms: 4-[4-(prop-1-en-1-yl)cyclohexyl]benzonitrile
Molecular Weight (g/mol): 225.34
Molecular Formula: C16H19N
InChi Key: WFVBLRKVRNUULX-UHFFFAOYSA-N
SMILES: CC=CC1CCC(CC1)C1=CC=C(C=C1)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results