accessibility menu, dialog, popup

UserName

CAS RN 96020-91-6

HH2OClNH2NH2FFOHO

CAS RN 96020-91-6

IUPAC Name: hydrogen 2,5-diamino-2-(difluoromethyl)pentanoic acid hydrate chloride
Synonyms: hydrogen vaniqa hydrate chloride
Molecular Weight (g/mol): 236.64
Molecular Formula: C6H15ClF2N2O3
InChi Key: FJPAMFNRCFEGSD-UHFFFAOYNA-N
SMILES: [H+].O.[Cl-].NCCCC(N)(C(F)F)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results