accessibility menu, dialog, popup

UserName

CAS RN 95-25-0

ClOOHN

CAS RN 95-25-0

IUPAC Name: 5-chloro-2,3-dihydro-1,3-benzoxazol-2-one
Synonyms: chlorzoxazone chlorzoxazon 氯唑沙宗 chloroxazone chlorzoxazone (inn) neoflex
Molecular Weight (g/mol): 169.56
Molecular Formula: C7H4ClNO2
InChi Key: TZFWDZFKRBELIQ-UHFFFAOYSA-N
SMILES: ClC1=CC=C2OC(=O)NC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)