accessibility menu, dialog, popup

UserName

CAS RN 94544-77-1

NNBrBr

CAS RN 94544-77-1

IUPAC Name: 5,8-dibromo-2,3-diphenylquinoxaline
Synonyms: 5,8-dibromo-2,3-diphenylquinoxaline
Molecular Weight (g/mol): 440.14
Molecular Formula: C20H12Br2N2
InChi Key: LELMSURPXIAXOW-UHFFFAOYSA-N
SMILES: BrC1=CC=C(Br)C2=NC(C3=CC=CC=C3)=C(N=C12)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results