CAS RN 944451-28-9
IUPAC Name:
1-methyl-4-(propan-2-yl)benzene; dichlororuthenium; {4-[5-(diphenylphosphanyl)-2H-1,3-benzodioxol-4-yl]-2H-1,3-benzodioxol-5-yl}diphenylphosphane
Synonyms:
cymene; dichlororuthenium; segphos
Molecular Weight (g/mol):
916.78
Molecular Formula:
C48H42Cl2O4P2Ru
InChi Key:
HZLSGDCLBWZKRB-UHFFFAOYSA-L
SMILES:
Cl[Ru]Cl.CC(C)C1=CC=C(C)C=C1.C1OC2=C(O1)C(=C(C=C2)P(C1=CC=CC=C1)C1=CC=CC=C1)C1=C(C=CC2=C1OCO2)P(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
[RuCl(p-cymene)((R)-segphos(regR))]Cl, TCI America™
CAS: 944451-28-9 Molecular Formula: C48H42Cl2O4P2Ru Molecular Weight (g/mol): 916.778 MDL Number: MFCD09753017 InChI Key: HZLSGDCLBWZKRB-UHFFFAOYSA-L Synonym: Chloro[(R)-(+)-5,5′C-bis(diphenylphosphino)-4,4′C-bi-1,3-benzodioxole](p-cymene)ruthenium(II) Chloride PubChem CID: 72376325 IUPAC Name: dichlororuthenium;[4-(5-diphenylphosphanyl-1,3-benzodioxol-4-yl)-1,3-benzodioxol-5-yl]-diphenylphosphane;1-methyl-4-propan-2-ylbenzene SMILES: CC1=CC=C(C=C1)C(C)C.C1OC2=C(O1)C(=C(C=C2)P(C3=CC=CC=C3)C4=CC=CC=C4)C5=C(C=CC6=C5OCO6)P(C7=CC=CC=C7)C8=CC=CC=C8.Cl[Ru]Cl