CAS RN 939-52-6
IUPAC Name:
4-chloro-1-phenylbutan-1-one
Synonyms:
4-chloro-1-phenylbutan-1-one
Molecular Weight (g/mol):
182.65
Molecular Formula:
C10H11ClO
InChi Key:
GHEFQKHLHFXSBR-UHFFFAOYSA-N
SMILES:
ClCCCC(=O)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
4-Chlorobutyrophenone, 95%, Thermo Scientific™
4-Chlorobutyrophenone, 95%, Thermo Scientific™
CAS: 939-52-6 Molecular Formula: C10H11ClO Molecular Weight (g/mol): 182.65 MDL Number: MFCD00001005 InChI Key: GHEFQKHLHFXSBR-UHFFFAOYSA-N Synonym: 4-chlorobutyrophenone,gamma-chlorobutyrophenone,4-chloro-1-oxo-1-phenylbutane,.gamma.-chlorobutyrophenone,1-butanone,4-chloro-1-phenyl,chlorpropylphenylketon,4-chlorobutyrylbenzene,4-chlorobutyro-phenone,3-benzoylpropyl chloride,.omega.-chlorobutyrophenone PubChem CID: 253533 IUPAC Name: 4-chloro-1-phenylbutan-1-one SMILES: ClCCCC(=O)C1=CC=CC=C1