accessibility menu, dialog, popup

UserName

CHCH

CAS RN 935-14-8

IUPAC Name: 1,4-diethynylbenzene
Synonyms: 1,4-diethynylbenzene
Molecular Weight (g/mol): 126.16
Molecular Formula: C10H6
InChi Key: MVLGANVFCMOJHR-UHFFFAOYSA-N
SMILES: C#CC1=CC=C(C=C1)C#C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)