accessibility menu, dialog, popup

UserName

CH3H3CO(R)(R)OOHHO

CAS RN 93379-48-7

IUPAC Name: [(4R,5R)-5-(hydroxydiphenylmethyl)-2,2-dimethyl-1,3-dioxolan-4-yl]diphenylmethanol
Synonyms: [(4R,5R)-5-(hydroxydiphenylmethyl)-2,2-dimethyl-1,3-dioxolan-4-yl]diphenylmethanol
Molecular Weight (g/mol): 466.58
Molecular Formula: C31H30O4
InChi Key: OWVIRVJQDVCGQX-VSGBNLITSA-N
SMILES: CC1(C)O[C@H]([C@@H](O1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results