accessibility menu, dialog, popup

UserName

CAS RN 932-17-2

H3CONO

CAS RN 932-17-2

IUPAC Name: 1-acetylpyrrolidin-2-one
Synonyms: 2-pyrrolidinone, 1-acetyl-
Molecular Weight (g/mol): 127.14
Molecular Formula: C6H9NO2
InChi Key: YLHUPYSUKYAIBW-UHFFFAOYSA-N
SMILES: CC(=O)N1CCCC1=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results