accessibility menu, dialog, popup

UserName

H3CSiClClCl

CAS RN 928-65-4

IUPAC Name: trichloro(hexyl)silane
Synonyms: hexyltrichlorosilane
Molecular Weight (g/mol): 219.61
Molecular Formula: C6H13Cl3Si
InChi Key: LFXJGGDONSCPOF-UHFFFAOYSA-N
SMILES: CCCCCC[Si](Cl)(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results