accessibility menu, dialog, popup

UserName

CAS RN 927-62-8

H3CNHCH3CH3

CAS RN 927-62-8

IUPAC Name: butyldimethylazanium
Synonyms: butyldimethylazanium
Molecular Weight (g/mol): 102.2
Molecular Formula: C6H16N
InChi Key: DJEQZVQFEPKLOY-UHFFFAOYSA-O
SMILES: CCCC[NH+](C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results