accessibility menu, dialog, popup

UserName

CH3OOCH3(E/Z)

CAS RN 92666-21-2

IUPAC Name: 1-(benzyloxy)-2-methoxy-4-(prop-1-en-1-yl)benzene
Synonyms: 1-(benzyloxy)-2-methoxy-4-(prop-1-en-1-yl)benzene
Molecular Weight (g/mol): 254.33
Molecular Formula: C17H18O2
InChi Key: YKSSSKBJDZDZTD-UHFFFAOYSA-N
SMILES: COC1=C(OCC2=CC=CC=C2)C=CC(C=CC)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results